4-((tert-Butyldimethylsilyl)oxy)cyclohexanol 95%

Supplier: Ambeed
Warning

A743805-1G A743805-250MG A743805-5G
77036-062EA 101.85 USD
77036-062 77036-064 77036-066
4-((tert-Butyldimethylsilyl)oxy)cyclohexanol 95%
4-((tert-Butyldimethylsilyl)oxy)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₂₆O₂Si
MW: 230.41 g/mol
MDL Number: MFCD13190776
CAS Number: 126931-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR