N2-Boc-L-arginine hydrochloride hydrate 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Arg-OH·HCl·H2O, Boc-L-Arg-OH.HCl hydrate

A743595-100G A743595-25G A743595-500G
77006-214EA 45.62 USD
77006-214 77006-216 77006-218
N2-Boc-L-arginine hydrochloride hydrate 98%
N2-Boc-L-arginine hydrochloride hydrate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₄O₄
MW: 328.79 g/mol
MDL Number: MFCD00065552
CAS Number: 114622-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR