4,5-Difluoro-m-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluoro-5-methylbenzoic acid

A743089-100MG A743089-250MG
77018-982EA 35 USD
77018-982 77018-984
4,5-Difluoro-m-toluic acid 98%
4,5-Difluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD09832332
CAS Number: 1017778-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR