(Boc-Cys-OH)₂ 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-di-Boc-L-cystine, AK-32714, (Boc-Cys-OH)2, Bis-Boc-L-cystine, (Boc-Cystine-OH)₂

A742726-500G A742726-25G A742726-100G A742726-5G
77036-060EA 441 USD
77036-060 77573-380 77882-628 77882-629
(Boc-Cys-OH)₂ 98%
(Boc-Cys-OH)₂

Precaución: for Research Use Only

Formula: C₁₆H₂₈N₂O₈S₂
MW: 440.526 g/mol
MDL Number: MFCD00038250
CAS Number: 10389-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR