1-Bromo-9H-fluoren-9-one 95%
Supplier: Ambeed
|
Warning
|
A742625-100MG
A742625-1G
A742625-250MG
A742625-5G
77018-968EA
7
USD
77018-968
77018-970
77018-972
77018-974
1-Bromo-9H-fluoren-9-one 95%
1-Bromo-9H-fluoren-9-one
Precaución: for Research Use Only
Formula:
C₁₃H₇BrO |
MDL Number:
MFCD09966028 CAS Number: 36804-63-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...