2-Benzyloxy-6-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-(Benzyloxy)-6-fluorophenyl)boronic acid

A741637-100MG A741637-1G A741637-250MG A741637-5G
77006-196EA 16.98 USD
77006-196 77006-198 77006-200 77006-202
2-Benzyloxy-6-fluorophenylboronic acid 98%
2-Benzyloxy-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MW: 246.0420 g/mol
MDL Number: MFCD11617256
CAS Number: 1217500-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR