2,4,6-Triphenylaniline 97%

Supplier: Ambeed
Warning

Synonyms: 5'-Phenyl-[1,1':3',1''-terphenyl]-2'-amine

A741239-100MG A741239-1G A741239-250MG A741239-5G
77036-046EA 13.8 USD
77036-046 77036-048 77036-050 77036-052
2,4,6-Triphenylaniline 97%
2,4,6-Triphenylaniline

Precaución: for Research Use Only

Formula: C₂₄H₁₉N
MW: 321.42 g/mol
MDL Number: MFCD00012371
CAS Number: 6864-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR