Ethyl 6-chloro-1H-indazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A740733-100MG A740733-1G A740733-250MG
77018-952EA 24.41 USD
77018-952 77018-954 77018-956
Ethyl 6-chloro-1H-indazole-3-carboxylate 97%
Ethyl 6-chloro-1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MW: 224.64 g/mol
MDL Number: MFCD07371603
CAS Number: 885279-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR