(R)-3-Cyclohexylmorpholine 95+%

Supplier: Ambeed
Danger

A740567-100MG A740567-1G A740567-250MG
76992-958EA 56.23 USD
76992-958 76992-960 76992-962
(R)-3-Cyclohexylmorpholine 95+%
(R)-3-Cyclohexylmorpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO
MW: 169.26 g/mol
MDL Number: MFCD18683245
CAS Number: 1269969-36-9
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR