2-Nitro-6-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Danger

Synonyms: 2-Cyano-3-(trifluoromethyl)nitrobenzene, 2-Cyano-3-nitrobenzotrifluoride, 6-Nitro-α,α,α-trifluoro-o-tolunitrile

A739176-1G A739176-25G A739176-5G
76992-952EA 18.04 USD
76992-952 76992-954 76992-956
2-Nitro-6-(trifluoromethyl)benzonitrile 97%
2-Nitro-6-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₃N₂O₂
MW: 216.117 g/mol
MDL Number: MFCD16606330
CAS Number: 1227581-80-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR