(5-(Ethoxycarbonyl)pyridin-3-yl)boronic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 5-(Ethoxycarbonyl)pyridine-3-boronic acid

A738904-1G A738904-250MG A738904-5G
76992-946EA 67 USD
76992-946 76992-948 76992-950
(5-(Ethoxycarbonyl)pyridin-3-yl)boronic acid 95+%
(5-(Ethoxycarbonyl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₄
MDL Number: MFCD09952043
CAS Number: 875781-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR