alpha-(4-Biphenylyl)benzylamine 95%
Supplier: Ambeed
|
Warning
|
A738706-1G
A738706-250MG
77018-940EA
58
USD
77018-940
77614-274
alpha-(4-Biphenylyl)benzylamine 95%
alpha-(4-Biphenylyl)benzylamine
Precaución: for Research Use Only
Formula:
C₁₉H₁₇N MW: 259.34 g/mol |
MDL Number:
MFCD11848907 CAS Number: 91487-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...