4-(4-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid 95%

Supplier: Ambeed
Warning

A738555-100MG A738555-1G A738555-250MG
77018-932EA 39 USD
77018-932 77018-934 77018-936
4-(4-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid 95%
4-(4-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆BClO₃
MDL Number: MFCD06411357
CAS Number: 849062-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR