Risedronic acid sodium salt 98%

Supplier: Ambeed
Warning

A738541-1G A738541-5G A738541-100MG A738541-250MG A738541-25G
77006-194EA 23 USD
77006-194 77602-894 77674-028 77674-030 77674-032
Risedronic acid sodium salt 98%
Risedronic acid sodium salt

Precaución: for Research Use Only

Formula: C₇H₁₀NNaO₇P₂
MDL Number: MFCD01706268
CAS Number: 115436-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR