2-(2-[3-(S)-(3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl)-3-hydroxy-n-propyl]phenyl)-2-propanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(3-(2-(7-Chloroquinolin-2-yl)vinyl)phenyl)-3-(2-(2-hydroxypropan-2-yl)phenyl)propan-1-ol

A738470-100G A738470-1G A738470-25G A738470-5G
77006-186EA 73 USD
77006-186 77006-188 77006-190 77006-192
2-(2-[3-(S)-(3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl)-3-hydroxy-n-propyl]phenyl)-2-propanol 97%
2-(2-[3-(S)-(3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl)-3-hydroxy-n-propyl]phenyl)-2-propanol

Precaución: for Research Use Only

Formula: C₂₉H₂₈ClNO₂
MDL Number: MFCD08460100
CAS Number: 142569-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR