5,10,15,20-Tetrakis(p-tolyl)porphyrin 98%

Supplier: Ambeed
Warning

Synonyms: 5,10,15,20-Tetrakis(4-methylphenyl)porphyrin, 5,10,15,20-Tetrakis(4-methylphenyl)-21H,23H-porphine, 5,10,15,20-Tetrakis(p-tolyl)-21H,23H-porphine

A738180-1G A738180-250MG A738180-5G
77018-926EA 43 USD
77018-926 77018-928 77018-930
5,10,15,20-Tetrakis(p-tolyl)porphyrin 98%
5,10,15,20-Tetrakis(p-tolyl)porphyrin

Precaución: for Research Use Only

Formula: C₄₈H₃₈N₄
MDL Number: MFCD00239499
CAS Number: 14527-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR