4-Boc-1-Oxa-4,9-diazaspiro[5.5]undecane 95%

Supplier: Ambeed
Warning

A738020-100MG A738020-1G A738020-250MG A738020-5G
77036-040EA 34 USD
77036-040 77036-042 77036-044 77873-165
4-Boc-1-Oxa-4,9-diazaspiro[5.5]undecane 95%
4-Boc-1-Oxa-4,9-diazaspiro[5.5]undecane

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₃
MDL Number: MFCD11227066
CAS Number: 1023595-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR