Boc-3-(1-naphthyl)-L-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-1-Nal-OH, N-Boc-3-(1-naphthyl)-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)-3-(naphthalen-1-yl)propanoic acid

A737939-1G A737939-250MG A737939-25G A737939-5G
77036-032EA 33.95 USD
77036-032 77036-034 77036-036 77036-038
Boc-3-(1-naphthyl)-L-alanine 98%
Boc-3-(1-naphthyl)-L-alanine

Precaución: for Research Use Only

Formula: C₁₈H₂₁NO₄
MW: 315.369 g/mol
MDL Number: MFCD00085377
CAS Number: 55447-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR