(1-Methyl-1H-indol-6-yl)methanol 98%
Supplier: Ambeed
|
Warning
|
A737196-1G
A737196-250MG
A737196-5G
77036-024EA
93.36
USD
77036-024
77036-026
77036-028
(1-Methyl-1H-indol-6-yl)methanol 98%
(1-Methyl-1H-indol-6-yl)methanol
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO |
MDL Number:
MFCD08690252 CAS Number: 199590-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...