Methyl 4-amino-1-methyl-1H-pyrazole-5-carboxylate 98%
Supplier: Ambeed
|
Danger
|
A736047-1G
A736047-5G
A736047-250MG
76992-940EA
62.6
USD
76992-940
77573-368
77684-660
Methyl 4-amino-1-methyl-1H-pyrazole-5-carboxylate 98%
Methyl 4-amino-1-methyl-1H-pyrazole-5-carboxylate
Precaución: For research use only.
Formula:
C₆H₉N₃O₂ MW: 155.16 g/mol |
MDL Number:
MFCD04969992 CAS Number: 923283-54-9 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...