4-Borono-3-fluorothioanisole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-methylthiophenylboronic acid, (2-Fluoro-4-(methylthio)phenyl)boronic acid, 2-Fluoro-4-(methylsulphanyl)benzeneboronic acid

A735257-100MG A735257-1G A735257-250MG A735257-5G
76992-934EA 11 USD
76992-934 76992-936 76992-938 77891-452
4-Borono-3-fluorothioanisole 98%
4-Borono-3-fluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₈BFO₂S
MDL Number: MFCD09027247
CAS Number: 957060-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR