7-Methylquinoline-8-carboxylic acid 95%

Supplier: Ambeed
Warning

A734945-1G A734945-250MG
77018-914EA 422 USD
77018-914 77018-916
7-Methylquinoline-8-carboxylic acid 95%
7-Methylquinoline-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MDL Number: MFCD13192614
CAS Number: 70585-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR