5-(Methoxycarbonyl)pyridine-3-boronic acid, pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate, 3-(Methoxycarbonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 5-(Methoxycarbonyl)pyridine-3-boronic acid pinacol ester

A734865-1G A734865-5G A734865-250MG A734865-25G
77036-020EA 30 USD
77036-020 77036-022 77669-978 77873-160
5-(Methoxycarbonyl)pyridine-3-boronic acid, pinacol ester 95%
5-(Methoxycarbonyl)pyridine-3-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₄
MW: 263.097 g/mol
MDL Number: MFCD09027079
CAS Number: 1025718-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR