3-(3-Bromo-2-fluorophenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A734654-100MG A734654-10G A734654-1G A734654-5G
77036-012EA 7 USD
77036-012 77036-014 77036-016 77036-018
3-(3-Bromo-2-fluorophenyl)propanoic acid 95%
3-(3-Bromo-2-fluorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MDL Number: MFCD18391365
CAS Number: 1261814-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR