(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanol ≥95%, ee 99%
Supplier: Ambeed
|
Warning
|
A734508-1G
A734508-250MG
A734508-25G
A734508-5G
76992-926EA
18
USD
76992-926
76992-928
76992-930
76992-932
(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanol ≥95%, ee 99%
(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetra(1-naphthyl)-1,3-dioxolane-4,5-dimethanol
Precaución: for Research Use Only
Formula:
C₄₇H₃₈O₄ |
MDL Number:
MFCD00192091 CAS Number: 137536-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...