N-Boc-3-(methylamino)-1-propanol 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-N-boc-aminopropan-3-ol

A734482-250MG A734482-5G A734482-1G A734482-25G A734482-500G
77036-008EA 6 USD
77036-008 77036-010 77595-154 77674-010 77674-012
N-Boc-3-(methylamino)-1-propanol 98%
N-Boc-3-(methylamino)-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MDL Number: MFCD09751790
CAS Number: 98642-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR