1-Methyl-1H-indazole-6-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-indazole-6-boronic Acid, 1-Methylindazole-6-boronic acid

A734036-10G A734036-1G A734036-5G A734036-250MG
77036-002EA 197 USD
77036-002 77036-004 77036-006 77573-364
1-Methyl-1H-indazole-6-boronic acid 95%
1-Methyl-1H-indazole-6-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD09870053
CAS Number: 1150114-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR