4-Amino-1-methyl-1H-pyrazole-3-carboxamide 95%

Supplier: Ambeed
Warning

A733903-100MG A733903-250MG A733903-50MG
77035-996EA 59.42 USD
77035-996 77035-998 77036-000
4-Amino-1-methyl-1H-pyrazole-3-carboxamide 95%
4-Amino-1-methyl-1H-pyrazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₅H₈N₄O
MDL Number: MFCD03422546
CAS Number: 3920-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR