Sodium-2-naphthol-7-sulfonate hydrate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Naphthol-7-sulphonic acid Sodium Salt, F acid Sodium Salt, Cassella's acid Sodium Salt, Sodium 2-naphthol-7-sulphonate, 7-Hydroxy-2-naphthalenesulphonic acid Sodium Salt, Sodium 7-Hydroxy-2-naphthalenesulphonate

A733677-100G A733677-25G A733677-5G
77006-174EA 55.17 USD
77006-174 77006-176 77006-178
Sodium-2-naphthol-7-sulfonate hydrate 98%
Sodium-2-naphthol-7-sulfonate hydrate

Precaución: for Research Use Only

Formula: C₁₀H₈O₄S
MW: 246.21 g/mol
MDL Number: MFCD00021625
CAS Number: 135-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR