2,3,4,6-Tetra-O-acetyl-D-glucopyranose 95%

Supplier: Ambeed
Warning

A733645-100MG A733645-10G A733645-1G A733645-250MG A733645-5G A733645-25G
77035-986EA 6 USD
77035-986 77035-988 77035-990 77035-992 77035-994 77573-358
2,3,4,6-Tetra-O-acetyl-D-glucopyranose 95%
2,3,4,6-Tetra-O-acetyl-D-glucopyranose

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₁₀
MDL Number: MFCD00061634
CAS Number: 10343-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR