3-(Benzyloxy)-5-chloroaniline ≥98%

Supplier: Ambeed
Warning

A732876-5G A732876-1G A732876-250MG
77035-984EA 355 USD
77035-984 77873-154 77873-155
3-(Benzyloxy)-5-chloroaniline ≥98%
3-(Benzyloxy)-5-chloroaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD11655923
CAS Number: 1100752-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR