4-Ethoxy-2,3-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A732528-100MG A732528-1G A732528-250MG
76992-920EA 33.95 USD
76992-920 76992-922 76992-924
4-Ethoxy-2,3-difluorobenzaldehyde 98%
4-Ethoxy-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
MDL Number: MFCD09258700
CAS Number: 126162-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR