4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3-trifluoromethylphenylboronic acid pinacol ester

A732092-250MG A732092-1G
76992-918EA 63.66 USD
76992-918 77573-354
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol 98%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₆BF₃O₃
MW: 288.07 g/mol
MDL Number: MFCD16994250
CAS Number: 1243143-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR