3-Cyano-1H-pyrrole-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A731930-100MG
A731930-250MG
76992-914EA
190.97
USD
76992-914
76992-916
3-Cyano-1H-pyrrole-2-carboxylic acid 95%
3-Cyano-1H-pyrrole-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₄N₂O₂ MW: 136.11 g/mol |
MDL Number:
MFCD19221270 CAS Number: 7126-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...