5-Bromo-2-methylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A729412-100MG A729412-250MG A729412-1G
77018-894EA 36 USD
77018-894 77018-896 77573-340
5-Bromo-2-methylpyrimidin-4-amine 95%
5-Bromo-2-methylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MDL Number: MFCD00223699
CAS Number: 24343-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR