(3-Fluoro-5-methylphenyl)acetic acid 97%

Supplier: Ambeed
Warning

A729396-10G A729396-1G A729396-250MG A729396-25G A729396-5G
77035-970EA 251 USD
77035-970 77035-972 77035-974 77035-976 77035-978
(3-Fluoro-5-methylphenyl)acetic acid 97%
(3-Fluoro-5-methylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD03094318
CAS Number: 518070-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR