(2S,4S)-tert-Butyl 4-amino-2-(hydroxymethyl)pyrrolidine-1-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A728364-100MG A728364-250MG
76992-898EA 56.23 USD
76992-898 76992-900
(2S,4S)-tert-Butyl 4-amino-2-(hydroxymethyl)pyrrolidine-1-carboxylate hydrochloride 98%
(2S,4S)-tert-Butyl 4-amino-2-(hydroxymethyl)pyrrolidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁ClN₂O₃
MW: 252.73 g/mol
MDL Number: MFCD10568358
CAS Number: 1279037-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR