Methyl-3-formylindole-6-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Formylindole-6-carboxylate

A727410-10G A727410-1G A727410-250MG A727410-5G
77006-154EA 167.63 USD
77006-154 77006-156 77006-158 77006-160
Methyl-3-formylindole-6-carboxylate 98%
Methyl-3-formylindole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.197 g/mol
MDL Number: MFCD00216479
CAS Number: 133831-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR