4'-Chlorochalcone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Chlorophenyl)-3-phenylprop-2-en-1-one, 4'-Chlorobenzylidenacetophenone

A726319-5G A726319-25G A726319-100G A726319-1G
76992-896EA 7 USD
76992-896 77891-434 77891-432 77891-433
4'-Chlorochalcone 98%
4'-Chlorochalcone

Precaución: for Research Use Only

Formula: C₁₅H₁₁ClO
MDL Number: MFCD00016343
CAS Number: 956-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR