N4-Benzoylcytidine 98%

Supplier: Ambeed
Warning

Synonyms: N-Benzoylcytidine

A725963-100G A725963-1G A725963-25G A725963-5G
77035-924EA 300.24 USD
77035-924 77035-926 77035-928 77035-930
N4-Benzoylcytidine 98%
N4-Benzoylcytidine

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₆
MW: 347.33 g/mol
MDL Number: MFCD00010572
CAS Number: 13089-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR