4,4'-Diamino-2,2'-difluorobiphenyl 98%

Supplier: Ambeed
Warning

A725286-1G A725286-250MG A725286-5G
76992-886EA 145.35 USD
76992-886 76992-888 76992-890
4,4'-Diamino-2,2'-difluorobiphenyl 98%
4,4'-Diamino-2,2'-difluorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₁₀F₂N₂
MW: 220.21 g/mol
MDL Number: MFCD19302620
CAS Number: 316-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR