Chlorodifluoroacetic acid 98%

Supplier: Ambeed
Danger

Synonyms: Difluorochloroacetic acid

A724594-25G A724594-5G
77006-150EA 28 USD
77006-150 77006-152
Chlorodifluoroacetic acid 98%
Chlorodifluoroacetic acid

Precaución: for Research Use Only

Formula: C₂HClF₂O₂
MW: 130.48 g/mol
Boiling Pt: 122
MDL Number: MFCD00004176
CAS Number: 76-04-0
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR