Ethyl benzo[d]thiazole-6-carboxylate 97%

Supplier: Ambeed
Warning

A724254-100MG A724254-1G A724254-250MG A724254-5G
77035-908EA 109.28 USD
77035-908 77035-910 77035-912 77035-914
Ethyl benzo[d]thiazole-6-carboxylate 97%
Ethyl benzo[d]thiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂S
MW: 207.24 g/mol
MDL Number: MFCD02089769
CAS Number: 19989-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR