4-[(Trifluoroacetyl)amino]benzoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(2,2,2-Trifluoroacetamido)benzoic acid
A724063-1G
A724063-250MG
A724063-5G
77035-902EA
108.22
USD
77035-902
77035-904
77035-906
4-[(Trifluoroacetyl)amino]benzoic acid 95%
4-[(Trifluoroacetyl)amino]benzoic acid
Precaución: for Research Use Only
Formula:
C₉H₆F₃NO₃ MW: 233.15 g/mol |
MDL Number:
MFCD00454055 CAS Number: 404-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...