1-Fluoro-2,5-dimethyl-4-nitrobenzene 95%

Supplier: Ambeed
Danger

A723883-100MG A723883-1G A723883-250MG A723883-5G
76992-866EA 48.81 USD
76992-866 76992-868 76992-870 76992-872
1-Fluoro-2,5-dimethyl-4-nitrobenzene 95%
1-Fluoro-2,5-dimethyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.15 g/mol
MDL Number: MFCD28049404
CAS Number: 1736-88-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR