3-tert-Butylbenzaldehyde 98%

Supplier: Ambeed
Warning

A722883-250MG A722883-5G A722883-1G
77035-900EA 18.04 USD
77035-900 77552-234 77573-328
3-tert-Butylbenzaldehyde 98%
3-tert-Butylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MDL Number: MFCD11100027
CAS Number: 23039-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR