S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulfonate 99+%

Supplier: Ambeed
Warning

Synonyms: KB-R 7943, 4-((4-Nitrobenzyl)oxy)phenethyl carbamimidothioate methanesulphonate, Carbamimidothioic acid 4-[(4-Nitrobenzyl)oxy]phenethyl Ester Methanesulphonate, 4-[(4-Nitrobenzyl)oxy]phenethyl Carbamimidothioate Methanesulphonate

A722708-10MG A722708-1MG A722708-50MG A722708-5MG
77035-892EA 71.09 USD
77035-892 77035-894 77035-896 77035-898
S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulfonate 99+%
S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulfonate

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₃S
MW: 427.49 g/mol
MDL Number: MFCD00952138
CAS Number: 182004-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR