4,4'-Diaminodiphenyl ether 98%

Supplier: Ambeed
Danger

Synonyms: p-Aminophenyl ether4-Aminophenyl ether4,4-Oxydianiline4,4'-OxydianilineBis(4-aminophenyl) ether4,4'-Oxydianiline zone-refined

A722543-25G
77035-890EA 5.15 USD
77035-890
4,4'-Diaminodiphenyl ether 98%
4,4'-Diaminodiphenyl ether

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
MDL Number: MFCD00007863
CAS Number: 101-80-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR