Methyl 3-(2-fluorophenyl)-3-oxopropanoate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A722004-5G A722004-25G
77018-868EA 40 USD
77018-868 77684-638
Methyl 3-(2-fluorophenyl)-3-oxopropanoate 95%, Ambeed.Inc
Methyl 3-(2-fluorophenyl)-3-oxopropanoate

Precaución: For research use only.

Formula: C₁₀H₉FO₃
MW: 196.175 g/mol
MDL Number: MFCD03424763
CAS Number: 185302-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR