(8S)-5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: (S)-THQ-amine dihydrochloride, (S)-5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride

A721518-250MG A721518-100MG
77018-866EA 65 USD
77018-866 77614-222
(8S)-5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride 97%
(8S)-5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂N₂·2ClH
MDL Number: MFCD17926204
CAS Number: 865303-57-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR